Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_030651.3 | NP_085154.3 | 4 | 895 | |
GenBank mRNA | AK054885.1 | - | 4 | 895 | |
AK123424.1 | - | 3 | 1777 | ||
BC013201.1 | - | 6 | 721 | 3924575 | |
BC063046.1 | - | 4 | 843 | 5197474 |