Gene Transcripts
- Gene Symbol
- TTTY9A
- Entrez Gene ID
- 83864
- Gene Name
- testis-specific transcript, Y-linked 9A (non-protein coding)
- Gene Aliases
- NCRNA00131, TTTY9, TTY9
- Chromosome Location
- Chr. Y: 18729882 - 18739197 on Build GRCh38
- UniGene ID
- Hs.536002
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_001530.1 |
|
5 |
1437 |
|
- Gene Symbol
- TTTY9B
- Entrez Gene ID
- 425057
- Gene Name
- testis-specific transcript, Y-linked 9B (non-protein coding)
- Gene Aliases
- NCRNA00132, TTTY9, TTTY9A, TTY9
- Chromosome Location
- Chr. Y: 18581206 - 18590521 on Build GRCh38
- UniGene ID
- Hs.724282
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002159.1 |
|
5 |
1437 |
|
View Full Product Details