Gene Transcripts

Gene Symbol
CACNB2
Entrez Gene ID
783
Gene Name
calcium voltage-gated channel auxiliary subunit beta 2
Gene Aliases
CACNLB2, CAVB2, MYSB
Chromosome Location
Chr. 10: 18140560 - 18545881 on Build GRCh38
UniGene ID
Hs.59093
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000724.3 NP_000715.2 13 4375
NM_001167945.1 NP_001161417.1 14 4094
NM_201570.2 NP_963864.1 13 4229
NM_201571.3 NP_963865.2 14 4208
NM_201572.3 NP_963866.2 14 4136
NM_201590.2 NP_963884.2 13 4005
NM_201593.2 NP_963887.2 14 3871
NM_201596.2 NP_963890.2 14 3985
NM_201597.2 NP_963891.1 14 3913
XM_005252588.3 XP_005252645.1 13 4119
XM_005252591.3 XP_005252648.1 8 3180
XM_006717502.3 XP_006717565.1 13 4038
XM_011519659.2 XP_011517961.1 13 3933
XM_011519660.2 XP_011517962.1 13 4476
XM_017016625.1 XP_016872114.1 14 4093
GenBank mRNA AB208917.1 - 13 4265

Gene Symbol
ARHGAP11B
Entrez Gene ID
89839
Gene Name
Rho GTPase activating protein 11B
Gene Aliases
B'-T, FAM7B1
Chromosome Location
Chr. 15: 30626676 - 30638810 on Build GRCh38
UniGene ID
Hs.659621
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK310319.1 - 9 939

Gene Symbol
CALN1
Entrez Gene ID
83698
Gene Name
calneuron 1
Gene Aliases
CABP8
Chromosome Location
Chr. 7: 71779491 - 72504271 on Build GRCh38
UniGene ID
Hs.592750
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001017440.2 NP_001017440.1 6 6000
NM_031468.3 NP_113656.2 7 5990
XM_011516594.2 XP_011514896.1 6 5842
XM_011516595.1 XP_011514897.1 6 5590
XM_011516596.2 XP_011514898.1 7 6485
XM_011516597.2 XP_011514899.1 6 6418
XM_011516599.1 XP_011514901.1 6 5789
XM_017012675.1 XP_016868164.1 8 6345
XM_017012676.1 XP_016868165.1 7 6127
XM_017012677.1 XP_016868166.1 6 5640
XM_017012678.1 XP_016868167.1 7 6102
XM_017012679.1 XP_016868168.1 6 6455
XM_017012680.1 XP_016868169.1 6 5754
XM_017012681.1 XP_016868170.1 6 5617
XM_017012682.1 XP_016868171.1 6 6167
XM_017012683.1 XP_016868172.1 7 5951
GenBank mRNA AF282250.1 - 7 5722
AK094469.1 - 1 1958

Gene Symbol
CARMN
Entrez Gene ID
728264
Gene Name
cardiac mesoderm enhancer-associated non-coding RNA
Gene Aliases
CARMEN, MIR143HG
Chromosome Location
Chr. 5: 149406845 - 149432836 on Build GRCh38
UniGene ID
Hs.519666
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BX640700.1 - 2 5084

Gene Symbol
CHAF1B
Entrez Gene ID
8208
Gene Name
chromatin assembly factor 1 subunit B
Gene Aliases
CAF-1, CAF-IP60, CAF1, CAF1A, CAF1P60, MPHOSPH7, MPP7
Chromosome Location
Chr. 21: 36380145 - 36416827 on Build GRCh38
UniGene ID
Hs.75238
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK092781.1 - 1 1416

Gene Symbol
COPS5
Entrez Gene ID
10987
Gene Name
COP9 signalosome subunit 5
Gene Aliases
CSN5, JAB1, MOV-34, SGN5
Chromosome Location
Chr. 8: 67043079 - 67062327 on Build GRCh38
UniGene ID
Hs.491912
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AB209134.1 - 6 3961

Gene Symbol
CROCCP2
Entrez Gene ID
84809
Gene Name
ciliary rootlet coiled-coil, rootletin pseudogene 2
Gene Aliases
CROCCL1
Chromosome Location
Chr. 1: 16618256 - 16630906 on Build GRCh38
UniGene ID
Hs.631865
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK090414.1 - 11 2030

Gene Symbol
EMC3
Entrez Gene ID
55831
Gene Name
ER membrane protein complex subunit 3
Gene Aliases
POB, TMEM111
Chromosome Location
Chr. 3: 9962680 - 10011248 on Build GRCh38
UniGene ID
Hs.475392
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK222953.1 - 7 1516

Gene Symbol
FAHD2A
Entrez Gene ID
51011
Gene Name
fumarylacetoacetate hydrolase domain containing 2A
Gene Aliases
CGI-105
Chromosome Location
Chr. 2: 95402691 - 95416702 on Build GRCh38
UniGene ID
Hs.531247
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_011511282.2 XP_011509584.1 1 23
XM_011511288.2 XP_011509590.1 1 23
XM_011511289.2 XP_011509591.1 1 23
XM_011511290.2 XP_011509592.1 1 23
XM_017004241.1 XP_016859730.1 1 23
XM_017004243.1 XP_016859732.1 1 23
XM_017004245.1 XP_016859734.1 1 23
XM_017004246.1 XP_016859735.1 1 23

Gene Symbol
HAT1
Entrez Gene ID
8520
Gene Name
histone acetyltransferase 1
Gene Aliases
KAT1
Chromosome Location
Chr. 2: 171922425 - 171983686 on Build GRCh38
UniGene ID
Hs.632532
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK127840.1 - 4 2367
AK309001.1 - 9 1260

Gene Symbol
IPO9-AS1
Entrez Gene ID
100873949
Gene Name
IPO9 antisense RNA 1
Gene Aliases
Chromosome Location
Chr. 1: 201688256 - 201829559 on Build GRCh38
UniGene ID
Hs.604842
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AI990057.1 - 1 29

Gene Symbol
ITGA8
Entrez Gene ID
8516
Gene Name
integrin subunit alpha 8
Gene Aliases
Chromosome Location
Chr. 10: 15513949 - 15720335 on Build GRCh38
UniGene ID
Hs.171311
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001291494.1 NP_001278423.1 29 6254
NM_003638.2 NP_003629.2 30 6299

Gene Symbol
KIAA1429
Entrez Gene ID
25962
Gene Name
KIAA1429
Gene Aliases
MSTP054, fSAP121
Chromosome Location
Chr. 8: 94487777 - 94553518 on Build GRCh38
UniGene ID
Hs.202238
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA CR627454.1 - 6 2332

Gene Symbol
LINC00581
Entrez Gene ID
100874531
Gene Name
long intergenic non-protein coding RNA 581
Gene Aliases
linc-MBOAT1-4
Chromosome Location
Chr. 6: 21486061 - 21511892 on Build GRCh38
UniGene ID
Hs.112750
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_103790.1 1 23
GenBank mRNA BX118339.1 - 1 27

Gene Symbol
LINC00910
Entrez Gene ID
100130581
Gene Name
long intergenic non-protein coding RNA 910
Gene Aliases
Chromosome Location
Chr. 17: 43369845 - 43388898 on Build GRCh38
UniGene ID
Hs.546897
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_027413.2 4 2460
GenBank mRNA AL832479.1 - 4 1874

Gene Symbol
LOC100996573
Entrez Gene ID
100996573
Gene Name
WAS/WASL-interacting protein family member 3-like
Gene Aliases
Chromosome Location
Chr. 12: 132550729 - 132559465 on Build GRCh38
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003846334.4 XP_003846382.2 1 4025

Gene Symbol
LOC101930105
Entrez Gene ID
101930105
Gene Name
zinc finger protein 839 pseudogene
Gene Aliases
Chromosome Location
Chr. X: 3854393 - 3886550 on Build GRCh38
UniGene ID
Hs.710994
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK097803.1 - 4 1505

Gene Symbol
NOC2L
Entrez Gene ID
26155
Gene Name
NOC2 like nucleolar associated transcriptional repressor
Gene Aliases
NET15, NET7, NIR, PPP1R112
Chromosome Location
Chr. 1: 944203 - 959299 on Build GRCh38
UniGene ID
Hs.405987
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK092491.1 - 1 194

Gene Symbol
NR1D2
Entrez Gene ID
9975
Gene Name
nuclear receptor subfamily 1 group D member 2
Gene Aliases
BD73, EAR-1R, RVR
Chromosome Location
Chr. 3: 23945260 - 23980618 on Build GRCh38
UniGene ID
Hs.37288
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AB209091.1 - 4 2105

Gene Symbol
NSUN4
Entrez Gene ID
387338
Gene Name
NOP2/Sun RNA methyltransferase family member 4
Gene Aliases
SHTAP
Chromosome Location
Chr. 1: 46340177 - 46365152 on Build GRCh38
UniGene ID
Hs.163424
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001256127.1 NP_001243056.1 6 4389
NM_001256128.1 NP_001243057.1 6 4021
NM_199044.3 NP_950245.2 6 4444
NR_045789.1 7 4619
NR_045790.1 5 4289
NR_045791.1 5 4107
GenBank mRNA BE542166.1 - 1 108
BM888153.1 - 1 117

Gene Symbol
NUDT9
Entrez Gene ID
53343
Gene Name
nudix hydrolase 9
Gene Aliases
NUDT10
Chromosome Location
Chr. 4: 87420002 - 87459454 on Build GRCh38
UniGene ID
Hs.149500
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_011532032.2 XP_011530334.1 1 1902

Gene Symbol
PDE12
Entrez Gene ID
201626
Gene Name
phosphodiesterase 12
Gene Aliases
2'-PDE, 2-PDE
Chromosome Location
Chr. 3: 57556247 - 57656480 on Build GRCh38
UniGene ID
Hs.572993
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001322177.1 NP_001309106.1 2 2666
NM_177966.6 NP_808881.3 3 2493
GenBank mRNA AK001526.1 - 1 762
AL831824.1 - 3 1997
BX647169.1 - 3 2459

Gene Symbol
POLH
Entrez Gene ID
5429
Gene Name
DNA polymerase eta
Gene Aliases
RAD30, RAD30A, XP-V, XPV
Chromosome Location
Chr. 6: 43576141 - 43620523 on Build GRCh38
UniGene ID
Hs.655467
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001291969.1 NP_001278898.1 9 6369
NM_001291970.1 NP_001278899.1 11 6657
NM_006502.2 NP_006493.1 11 6728

Gene Symbol
POLM
Entrez Gene ID
27434
Gene Name
DNA polymerase mu
Gene Aliases
Pol Mu, Tdt-N
Chromosome Location
Chr. 7: 44072062 - 44082540 on Build GRCh38
UniGene ID
Hs.596982
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_011515286.2 XP_011513588.1 6 2296
XM_017012004.1 XP_016867493.1 6 2280
XM_017012005.1 XP_016867494.1 7 2271

Gene Symbol
PTPN2
Entrez Gene ID
5771
Gene Name
protein tyrosine phosphatase, non-receptor type 2
Gene Aliases
PTN2, PTPT, TC-PTP, TCELLPTP, TCPTP
Chromosome Location
Chr. 18: 12785478 - 12884351 on Build GRCh38
UniGene ID
Hs.654527
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001308287.1 NP_001295216.1 8 2812
NM_002828.3 NP_002819.2 9 2917
XM_005258124.3 XP_005258181.1 10 2984
GenBank mRNA AB209569.1 - 8 2844
DB350067.1 - 1 191

Gene Symbol
RAD23B
Entrez Gene ID
5887
Gene Name
RAD23 homolog B, nucleotide excision repair protein
Gene Aliases
HHR23B, HR23B, P58
Chromosome Location
Chr. 9: 107283236 - 107332194 on Build GRCh38
UniGene ID
Hs.521640
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK293532.1 - 9 1618

Gene Symbol
RBBP5
Entrez Gene ID
5929
Gene Name
RB binding protein 5, histone lysine methyltransferase complex subunit
Gene Aliases
RBQ3, SWD1
Chromosome Location
Chr. 1: 205086142 - 205122022 on Build GRCh38
UniGene ID
Hs.519230
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001193272.1 NP_001180201.1 14 3031
NM_001193273.1 NP_001180202.1 14 3086
NM_005057.3 NP_005048.2 14 3145
XM_011509833.2 XP_011508135.1 14 3140
GenBank mRNA BC053856.1 - 14 3114
X85134.1 - 14 3108

Gene Symbol
RRP7A
Entrez Gene ID
27341
Gene Name
ribosomal RNA processing 7 homolog A
Gene Aliases
BK126B4.3, CGI-96
Chromosome Location
Chr. 22: 42508335 - 42519823 on Build GRCh38
UniGene ID
Hs.745006
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK092492.1 - 1 391

Gene Symbol
SETDB2
Entrez Gene ID
83852
Gene Name
SET domain bifurcated 2
Gene Aliases
C13orf4, CLLD8, CLLL8, KMT1F
Chromosome Location
Chr. 13: 49444293 - 49495003 on Build GRCh38
UniGene ID
Hs.631789
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AF277186.1 - 2 356

Gene Symbol
SLC25A29
Entrez Gene ID
123096
Gene Name
solute carrier family 25 member 29
Gene Aliases
C14orf69, CACL, ORNT3
Chromosome Location
Chr. 14: 100277986 - 100306547 on Build GRCh38
UniGene ID
Hs.578109
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017020980.1 XP_016876469.1 4 5116
XM_017020981.1 XP_016876470.1 3 4640
XM_017020982.1 XP_016876471.1 3 3170
XM_017020983.1 XP_016876472.1 3 3160
XM_017020984.1 XP_016876473.1 3 2874
XM_017020985.1 XP_016876474.1 4 2919
XM_017020986.1 XP_016876475.1 3 2355
XM_017020987.1 XP_016876476.1 4 2335
GenBank mRNA AK095532.1 - 1 282
AL832299.1 - 1 2337

Gene Symbol
SPACA9
Entrez Gene ID
11092
Gene Name
sperm acrosome associated 9
Gene Aliases
C9orf9, Mast
Chromosome Location
Chr. 9: 132878320 - 132890314 on Build GRCh38
UniGene ID
Hs.62595
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001316897.1 NP_001303826.1 4 1842
NM_001316898.1 NP_001303827.1 4 1835
NR_133631.1 3 1632
GenBank mRNA AK075259.1 - 4 1836
AK130389.1 - 1 1916
BC012940.1 - 4 1792 4478288

Gene Symbol
SPATA33
Entrez Gene ID
124045
Gene Name
spermatogenesis associated 33
Gene Aliases
C16orf55
Chromosome Location
Chr. 16: 89657744 - 89670458 on Build GRCh38
UniGene ID
Hs.655171
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC045670.1 - 2 1310

Gene Symbol
SRGAP3
Entrez Gene ID
9901
Gene Name
SLIT-ROBO Rho GTPase activating protein 3
Gene Aliases
ARHGAP14, MEGAP, SRGAP2, WRP
Chromosome Location
Chr. 3: 8980591 - 9362979 on Build GRCh38
UniGene ID
Hs.654743
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AB032982.1 - 9 5471

Gene Symbol
SYF2
Entrez Gene ID
25949
Gene Name
SYF2 pre-mRNA splicing factor
Gene Aliases
CBPIN, NTC31, P29, fSAP29
Chromosome Location
Chr. 1: 25222276 - 25232522 on Build GRCh38
UniGene ID
Hs.20013
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK303582.1 - 5 852

Gene Symbol
TMC2
Entrez Gene ID
117532
Gene Name
transmembrane channel like 2
Gene Aliases
C20orf145
Chromosome Location
Chr. 20: 2536607 - 2642185 on Build GRCh38
UniGene ID
Hs.352626
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK127751.1 - 4 3365

Gene Symbol
TMEM116
Entrez Gene ID
89894
Gene Name
transmembrane protein 116
Gene Aliases
Chromosome Location
Chr. 12: 111894936 - 112013219 on Build GRCh38
UniGene ID
Hs.506815
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AL833501.1 - 10 4748

Gene Symbol
USP9X
Entrez Gene ID
8239
Gene Name
ubiquitin specific peptidase 9, X-linked
Gene Aliases
DFFRX, FAF, FAM, MRX99, MRXS99F
Chromosome Location
Chr. X: 41085420 - 41236579 on Build GRCh38
UniGene ID
Hs.77578
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039590.2 NP_001034679.2 45 10590
NM_001039591.2 NP_001034680.2 45 10542
XM_005272675.4 XP_005272732.1 45 10820

Gene Symbol
WDSUB1
Entrez Gene ID
151525
Gene Name
WD repeat, sterile alpha motif and U-box domain containing 1
Gene Aliases
UBOX6, WDSAM1
Chromosome Location
Chr. 2: 159235793 - 159286799 on Build GRCh38
UniGene ID
Hs.20848
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017003481.1 XP_016858970.1 11 2014

Gene Symbol
ZMYM5
Entrez Gene ID
9205
Gene Name
zinc finger MYM-type containing 5
Gene Aliases
HSPC050, MYM, ZNF198L1, ZNF237
Chromosome Location
Chr. 13: 19823482 - 19863636 on Build GRCh38
UniGene ID
Hs.530988
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001142684.1 NP_001136156.1 8 2701
XM_005266593.3 XP_005266650.1 8 3088
XM_005266594.2 XP_005266651.1 7 2516
XM_006719894.2 XP_006719957.1 8 2811
XM_011535309.1 XP_011533611.1 8 3060
XM_011535310.1 XP_011533612.1 5 2514
XM_017020849.1 XP_016876338.1 4 2077
XM_017020850.1 XP_016876339.1 5 2144
GenBank mRNA BX647750.1 - 2 1366

Gene Symbol
ZNF614
Entrez Gene ID
80110
Gene Name
zinc finger protein 614
Gene Aliases
Chromosome Location
Chr. 19: 52013324 - 52028427 on Build GRCh38
UniGene ID
Hs.292336
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC004930.1 - 6 867

View Full Product Details