Gene Transcripts

Gene Symbol
TMEM263
Entrez Gene ID
90488
Gene Name
transmembrane protein 263
Gene Aliases
C12orf23
Chromosome Location
Chr. 12: 106955535 - 106974037 on Build GRCh38
UniGene ID
Hs.257664
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001319661.1 NP_001306590.1 3 453
NM_001319662.1 NP_001306591.1 3 474
NM_001319663.1 NP_001306592.1 5 619
NM_001319664.1 NP_001306593.1 5 637
NM_001319666.1 NP_001306595.1 5 640
NM_152261.3 NP_689474.1 4 521
XM_017020198.1 XP_016875687.1 4 708
GenBank mRNA AK001731.1 - 4 336
AK021783.1 - 2 136
AK022770.1 - 3 235
AK024021.1 - 4 348
BC020522.1 - 4 316 3916886
CX166555.1 - 5 513
DA609590.1 - 3 474
DB091654.1 - 4 521
DB226324.1 - 5 487
DB233011.1 - 5 442

View Full Product Details