Gene Transcripts

Gene Symbol
ST7-AS1
Entrez Gene ID
93653
Gene Name
ST7 antisense RNA 1
Gene Aliases
ST7AS1, ST7OT1
Chromosome Location
Chr. 7: 116952446 - 116954334 on Build GRCh38
UniGene ID
Hs.597516
Species
Homo sapiens
Interrogated Sequence Translated Protein RNADB ID Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_002330.1
1 1440
GenBank mRNA AF400039.1 - LIT2006
ST7OT1
1 1440

View Full Product Details