Gene Transcripts
- Gene Symbol
- TTTY4
- Entrez Gene ID
- 114761
- Gene Name
- testis-specific transcript, Y-linked 4 (non-protein coding)
- Gene Aliases
- LINC00123, NCRNA00123, TTTY4A, TTY4
- Chromosome Location
- Chr. Y: 22936455 - 22973284 on Build GRCh38
- UniGene ID
- Hs.679521
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_001525.1 |
|
4 |
1211 |
|
- Gene Symbol
- TTTY4B
- Entrez Gene ID
- 474149
- Gene Name
- testis-specific transcript, Y-linked 4B (non-protein coding)
- Gene Aliases
- LINC00124, NCRNA00124
- Chromosome Location
- Chr. Y: 24570202 - 24607025 on Build GRCh38
- UniGene ID
- Hs.679521
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002178.1 |
|
4 |
1211 |
|
- Gene Symbol
- TTTY4C
- Entrez Gene ID
- 474150
- Gene Name
- testis-specific transcript, Y-linked 4C (non-protein coding)
- Gene Aliases
- LINC00125, NCRNA00125
- Chromosome Location
- Chr. Y: 25063083 - 25099892 on Build GRCh38
- UniGene ID
- Hs.742604
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002177.1 |
|
4 |
1211 |
|
View Full Product Details