Gene Transcripts

Gene Symbol
GSE1
Entrez Gene ID
23199
Gene Name
Gse1 coiled-coil protein
Gene Aliases
CRHSP24, KIAA0182
Chromosome Location
Chr. 16: 85169512 - 85676206 on Build GRCh38
UniGene ID
Hs.461647
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_005255859.4 XP_005255916.3 1 1989
XM_005255860.3 XP_005255917.3 1 1996
XM_005255861.4 XP_005255918.3 1 1989
XM_017023084.1 XP_016878573.1 1 2161

View Full Product Details