Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_138573.3 | NP_612640.1 | 3 | 206 | |
XM_017021937.1 | XP_016877426.1 | 7 | 717 | ||
XM_017021938.1 | XP_016877427.1 | 6 | 2924 | ||
XM_017021939.1 | XP_016877428.1 | 6 | 486 | ||
XM_017021940.1 | XP_016877429.1 | 5 | 3262 | ||
XM_017021941.1 | XP_016877430.1 | 3 | 225 | ||
XM_017021942.1 | XP_016877431.1 | 3 | 243 | ||
XM_017021943.1 | XP_016877432.1 | 3 | 316 | ||
XM_017021944.1 | XP_016877433.1 | 6 | 3309 | ||
XM_017021945.1 | XP_016877434.1 | 6 | 3301 | ||
XM_017021946.1 | XP_016877435.1 | 6 | 3301 | ||
XM_017021947.1 | XP_016877436.1 | 6 | 3301 | ||
XM_017021948.1 | XP_016877437.1 | 4 | 3034 | ||
GenBank mRNA | AK307362.1 | - | 2 | 88 | |
AK311204.1 | - | 6 | 359 | ||
AK311507.1 | - | 3 | 212 | ||
AM392364.1 | - | 1 | 24 | ||
AM392365.1 | - | 1 | 24 | ||
BC017568.1 | - | 3 | 210 | 4071605 | |
BX538100.1 | - | 3 | 140 | ||
CR749519.1 | - | 3 | 201 |