Gene Transcripts

Gene Symbol
LINC01315
Entrez Gene ID
102723775
Gene Name
long intergenic non-protein coding RNA 1315
Gene Aliases
lnc-C22orf32-1
Chromosome Location
Chr. 22: 42364400 - 42369208 on Build GRCh38
UniGene ID
Hs.335413
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_120595.1 1 435
NR_120596.1 1 435
GenBank mRNA AI914073.1 - 2 389
AV709904.1 - 1 426
BC038245.1 - 1 401

View Full Product Details