Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001077191.1 | NP_001070659.1 | 2 | 1132 | |
NM_001077194.1 | NP_001070662.1 | 2 | 624 | ||
NM_001321950.1 | NP_001308879.1 | 2 | 998 | ||
NM_170699.2 | NP_733800.1 | 2 | 511 | ||
XM_011510743.1 | XP_011509045.1 | 2 | 1619 | ||
XM_017003467.1 | XP_016858956.1 | 3 | 1736 | ||
XM_017003468.1 | XP_016858957.1 | 3 | 2357 | ||
XM_017003469.1 | XP_016858958.1 | 3 | 1849 | ||
GenBank mRNA | AB086170.1 | - | 1 | 264 | |
AB089307.1 | - | 1 | 264 | ||
AK122658.1 | - | 2 | 998 | ||
AK122660.1 | - | 2 | 1132 | ||
BC033625.1 | - | 2 | 495 | 5221127 |