Gene Transcripts

Gene Symbol
RWDD4
Entrez Gene ID
201965
Gene Name
RWD domain containing 4
Gene Aliases
FAM28A, RWDD4A
Chromosome Location
Chr. 4: 183639635 - 183659219 on Build GRCh38
UniGene ID
Hs.133337
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001307922.1 NP_001294851.1 8 2066
NM_152682.3 NP_689895.2 8 2070
GenBank mRNA AK021702.1 - 8 1995
BC017472.1 - 8 2005 3909581

View Full Product Details