Gene Transcripts

Gene Symbol
SSX7
Entrez Gene ID
280658
Gene Name
SSX family member 7
Gene Aliases
Chromosome Location
Chr. X: 52644061 - 52654900 on Build GRCh38
UniGene ID
Hs.558712
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173358.2 NP_775494.1 6 576
XM_017029430.1 XP_016884919.1 5 417

View Full Product Details