Gene Transcripts

Gene Symbol
LOC285074
Entrez Gene ID
285074
Gene Name
anaphase promoting complex subunit 1 pseudogene
Gene Aliases
Chromosome Location
Chr. 2: 87030675 - 87076413 on Build GRCh38
UniGene ID
Hs.424184
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_026846.1 1 445
GenBank mRNA BC069048.1 - 1 227
BC132791.1 - 1 186
BC132793.1 - 1 186
BM454994.1 - 1 398
BX353812.2 - 1 445

Gene Symbol
RMND5A
Entrez Gene ID
64795
Gene Name
required for meiotic nuclear division 5 homolog A
Gene Aliases
CTLH, GID2, GID2A, RMD5, p44CTLH
Chromosome Location
Chr. 2: 86720291 - 86778041 on Build GRCh38
UniGene ID
Hs.75277
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022780.3 NP_073617.1 1 469
GenBank mRNA AK022815.1 - 1 446
BC012165.1 - 1 190 4636136
BC047668.1 - 1 399 6066798

View Full Product Details