Gene Transcripts

Gene Symbol
LUZP2
Entrez Gene ID
338645
Gene Name
leucine zipper protein 2
Gene Aliases
KFSP2566, PRO6246
Chromosome Location
Chr. 11: 24494304 - 25082640 on Build GRCh38
UniGene ID
Hs.144138
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009909.3 NP_001009909.2 2 433
NM_001252008.1 NP_001238937.1 2 429
NM_001252010.1 NP_001238939.1 2 433
XM_011520056.2 XP_011518358.1 2 3089
XM_017017648.1 XP_016873137.1 2 346
XM_017017649.1 XP_016873138.1 2 346
XM_017017650.1 XP_016873139.1 2 433
XM_017017651.1 XP_016873140.1 2 433
GenBank mRNA AK127695.1 - 2 433
AK309077.1 - 2 208
AL832641.3 - 2 345
AY164460.1 - 2 389
AY358386.1 - 2 264
BC132829.1 - 2 225 40146851
BC144177.1 - 2 221
DA706662.1 - 2 433

View Full Product Details