Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001102420.2 | NP_001095890.1 | 7 | 1257 | |
NM_001102421.2 | NP_001095891.1 | 6 | 1120 | ||
NM_001278243.1 | NP_001265172.1 | 7 | 836 | ||
NM_001278244.1 | NP_001265173.1 | 6 | 699 | ||
NM_001278245.1 | NP_001265174.1 | 7 | 833 | ||
NM_006007.3 | NP_005998.1 | 6 | 1117 | ||
GenBank mRNA | AF062346.1 | - | 6 | 840 | |
AF062347.1 | - | 6 | 695 | ||
AK025782.1 | - | 7 | 720 | ||
AK290849.1 | - | 6 | 840 | ||
AK307376.1 | - | 4 | 541 | ||
BC011018.1 | - | 6 | 956 | 4331933 | |
BC027707.2 | - | 7 | 1207 | 3945047 | |
BC073131.1 | - | 6 | 621 | 6137219 | |
BX649046.1 | - | 1 | 645 | ||
X75687.1 | - | 1 | 64 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NR_073180.1 | 2 | 134 | ||
GenBank mRNA | AK310497.1 | - | 3 | 712 | |
AK311181.1 | - | 2 | 108 | ||
BC037865.2 | - | 2 | 993 |