Gene Transcripts

Gene Symbol
ZNF644
Entrez Gene ID
84146
Gene Name
zinc finger protein 644
Gene Aliases
BM-005, MYP21, NatF, ZEP-2
Chromosome Location
Chr. 1: 90915298 - 91022269 on Build GRCh38
UniGene ID
Hs.173001
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_011542258.2 XP_011540560.1 11
GenBank mRNA DB001551.1 - 1 106

View Full Product Details