Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001142651.2 | NP_001136123.1 | 4 | 1511 | |
NM_001308177.1 | NP_001295106.1 | 3 | 965 | ||
NM_001308178.1 | NP_001295107.1 | 3 | 791 | ||
GenBank mRNA | GQ414757.1 | - | 4 | 1511 | |
GQ414758.1 | - | 3 | 965 | ||
GQ414759.1 | - | 3 | 791 |