Gene Transcripts

Gene Symbol
PIK3R3
Entrez Gene ID
8503
Gene Name
phosphoinositide-3-kinase regulatory subunit 3
Gene Aliases
p55, p55-GAMMA, p55PIK
Chromosome Location
Chr. 1: 46040140 - 46176495 on Build GRCh38
UniGene ID
Hs.655387
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001114172.1 NP_001107644.1 1 27
GenBank mRNA AK313726.1 - 1 39

View Full Product Details