Gene Transcripts

Gene Symbol
SERPINE2
Entrez Gene ID
5270
Gene Name
serpin family E member 2
Gene Aliases
GDN, GDNPF, PI-7, PI7, PN-1, PN1, PNI
Chromosome Location
Chr. 2: 223975048 - 224039319 on Build GRCh38
UniGene ID
Hs.38449
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001136530.1 NP_001130002.1 1 127
XM_005246641.2 XP_005246698.1 1 286
GenBank mRNA AK295564.1 - 1 95

View Full Product Details