Gene Transcripts
- Gene Symbol
- SLC37A3
- Entrez Gene ID
- 84255
- Gene Name
- solute carrier family 37 member 3
- Gene Aliases
- Chromosome Location
- Chr. 7: 140333752 - 140404352 on Build GRCh38
- UniGene ID
- Hs.446021
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
XM_011516626.2 |
XP_011514928.1
|
1, 2 |
139 |
|
XM_011516627.2 |
XP_011514929.1
|
1, 2 |
138 |
|
XM_017012712.1 |
XP_016868201.1
|
1, 2 |
115 |
|
XM_017012713.1 |
XP_016868202.1
|
1, 2 |
134 |
|
XM_017012714.1 |
XP_016868203.1
|
1, 2 |
155 |
|
XM_017012715.1 |
XP_016868204.1
|
1, 2 |
158 |
|
XM_017012716.1 |
XP_016868205.1
|
1, 2 |
162 |
|
XM_017012717.1 |
XP_016868206.1
|
1, 2 |
134 |
|
XM_017012718.1 |
XP_016868207.1
|
1, 2 |
133 |
|
GenBank mRNA |
AK131347.1 |
- |
1, 2 |
243 |
|
View Full Product Details