Gene Transcripts

Gene Symbol
DPRX
Entrez Gene ID
503834
Gene Name
divergent-paired related homeobox
Gene Aliases
Chromosome Location
Chr. 19: 53601257 - 53637057 on Build GRCh38
UniGene ID
Hs.579941
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012728.1 NP_001012746.1 1 64
XM_011527011.2 XP_011525313.1 2 124
XM_011527012.2 XP_011525314.1 2 134

View Full Product Details