Gene Transcripts
- Gene Symbol
- FKBP6
- Entrez Gene ID
- 8468
- Gene Name
- FK506 binding protein 6
- Gene Aliases
- FKBP36
- Chromosome Location
- Chr. 7: 73328152 - 73358637 on Build GRCh38
- UniGene ID
- Hs.661266
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001135211.2 |
NP_001128683.1
|
3, 4 |
359 |
|
NM_003602.4 |
NP_003593.3
|
3, 4 |
450 |
|
XM_006716153.2 |
XP_006716216.1
|
3, 4 |
354 |
|
XM_017012741.1 |
XP_016868230.1
|
3, 4 |
354 |
|
XM_017012742.1 |
XP_016868231.1
|
3, 4 |
357 |
|
GenBank mRNA |
AF038847.1 |
- |
3, 4 |
314 |
|
AK302121.1 |
- |
3, 4 |
347 |
|
AK310255.1 |
- |
3, 4 |
350 |
|
BC036817.1 |
- |
3, 4 |
322 |
5744865 |
BI461016.1 |
- |
3, 4 |
363 |
|
DB033483.1 |
- |
3, 4 |
385 |
|
DB450796.1 |
- |
3, 4 |
451 |
|
- Gene Symbol
- LOC541473
- Entrez Gene ID
- 541473
- Gene Name
- FK506 binding protein 6, 36kDa pseudogene
- Gene Aliases
- Chromosome Location
- Chr. 7: 75391949 - 75395461 on Build GRCh38
- UniGene ID
- Hs.645394
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003602.3 |
|
3, 4 |
433 |
|
GenBank mRNA |
BC022013.2 |
- |
3, 4 |
387 |
|
BC067783.1 |
- |
3, 4 |
406 |
|
View Full Product Details