Gene Transcripts

Gene Symbol
PRAMEF9
Entrez Gene ID
343070
Gene Name
PRAME family member 9
Gene Aliases
Chromosome Location
Chr. 1: 13175281 - 13179132 on Build GRCh38
UniGene ID
Hs.631873
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK124292.1 - 4 1155
BC101343.2 - 4 1141 40025151
BC137391.1 - 4 1141 9021389
BC171794.1 - 4 1141 9054448

Gene Symbol
PRAMEF11
Entrez Gene ID
440560
Gene Name
PRAME family member 11
Gene Aliases
Chromosome Location
Chr. 1: 12824380 - 12831410 on Build GRCh38
UniGene ID
Hs.551617
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001146344.2 NP_001139816.2 4 1132
XM_011541479.2 XP_011539781.1 3 1318
GenBank mRNA AL049680.1 - 4 1131
BC144426.1 - 4 1131 9052947

Gene Symbol
PRAMEF15
Entrez Gene ID
653619
Gene Name
PRAME family member 15
Gene Aliases
Chromosome Location
Chr. 1: 13315581 - 13322595 on Build GRCh38
UniGene ID
Hs.551617
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001098376.2 NP_001091846.1 4 1155

Gene Symbol
PRAMEF25
Entrez Gene ID
441873
Gene Name
PRAME family member 25
Gene Aliases
Chromosome Location
Chr. 1: 13073678 - 13078038 on Build GRCh38
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001310134.1 NP_001297063.1 3 1077

Gene Symbol
PRAMEF26
Entrez Gene ID
645359
Gene Name
PRAME family member 26
Gene Aliases
Chromosome Location
Chr. 1: 13148905 - 13153111 on Build GRCh38
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001306072.1 NP_001293001.1 3 1077

Gene Symbol
PRAMEF4
Entrez Gene ID
400735
Gene Name
PRAME family member 4
Gene Aliases
Chromosome Location
Chr. 1: 12879212 - 12886201 on Build GRCh38
UniGene ID
Hs.551617
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009611.4 NP_001009611.2 4 1132
GenBank mRNA AL049681.1 - 4 1132

Gene Symbol
PRAMEF5
Entrez Gene ID
343068
Gene Name
PRAME family member 5
Gene Aliases
PRAMEF23, PRAMEF5L
Chromosome Location
Chr. 1: 13254198 - 13263435 on Build GRCh38
UniGene ID
Hs.567864
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013407.2 NP_001013425.2 4 1155
GenBank mRNA BC101344.1 - 4 1141
BC130274.1 - 4 1140 40146305
BC130276.1 - 4 1140 40146307

Gene Symbol
PRAMEF6
Entrez Gene ID
440561
Gene Name
PRAME family member 6
Gene Aliases
Chromosome Location
Chr. 1: 12938472 - 12947580 on Build GRCh38
UniGene ID
Hs.567864
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001010889.2 NP_001010889.1 4 1141
GenBank mRNA BC101342.2 - 4 1141

View Full Product Details