Gene Transcripts

Gene Symbol
STAG3L3
Entrez Gene ID
442578
Gene Name
stromal antigen 3-like 3 (pseudogene)
Gene Aliases
STAG3L1, STAG3L2, STAG3L3P
Chromosome Location
Chr. 7: 72997295 - 73005943 on Build GRCh38
UniGene ID
Hs.632310
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_040582.2 1 96
GenBank mRNA AK316130.1 - 1 77
AY313779.1 - 1 68
BC108680.1 - 1 80
BG472630.1 - 1 38
DA505391.1 - 1 85
DA555040.1 - 1 96
DC405605.1 - 1 78

Gene Symbol
STAG3L1
Entrez Gene ID
54441
Gene Name
stromal antigen 3-like 1 (pseudogene)
Gene Aliases
STAG3L1P, STAG3L2, STAG3L3
Chromosome Location
Chr. 7: 75359200 - 75367809 on Build GRCh38
UniGene ID
Hs.632310
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_040583.2 1 80
GenBank mRNA AK289466.1 - 1 49
BG829469.1 - 1 80
DC419304.1 - 1 80

Gene Symbol
TRIM74
Entrez Gene ID
378108
Gene Name
tripartite motif containing 74
Gene Aliases
TRIM50C
Chromosome Location
Chr. 7: 72954845 - 73005932 on Build GRCh38
UniGene ID
Hs.645328
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017012197.1 XP_016867686.1 1 85

View Full Product Details