Gene Transcripts
- Gene Symbol
- TARP
- Entrez Gene ID
- 445347
- Gene Name
- TCR gamma alternate reading frame protein
- Gene Aliases
- CD3G, TCRG, TCRGC1, TCRGC2, TCRGV
- Chromosome Location
- Chr. 7: 38239577 - 38276741 on Build GRCh38
- UniGene ID
- Hs.534032
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001003799.1 |
NP_001003799.1
|
2, 3 |
389 |
|
NM_001003806.1 |
NP_001003806.1
|
2, 3 |
389 |
|
XM_011515407.2 |
XP_011513709.1
|
3, 4 |
609 |
|
XM_017012226.1 |
XP_016867715.1
|
1, 2 |
166 |
|
XM_017012227.1 |
XP_016867716.1
|
2, 3 |
599 |
|
XM_017012228.1 |
XP_016867717.1
|
1, 2 |
40 |
|
XM_017012229.1 |
XP_016867718.1
|
2, 3 |
811 |
|
XM_017012230.1 |
XP_016867719.1
|
2, 3 |
415 |
|
GenBank mRNA |
BC030554.1 |
- |
4, 5 |
1073 |
|
BC105589.1 |
- |
3, 4 |
607 |
4272944 |
BF674593.1 |
- |
2, 3 |
379 |
|
KC170727.1 |
- |
5, 6 |
746 |
|
M17325.1 |
- |
1, 2 |
308 |
|
M27334.1 |
- |
1, 2 |
329 |
|
X72500.1 |
- |
4, 5 |
755 |
|
- Gene Symbol
- TRGV9
- Entrez Gene ID
- 6983
- Gene Name
- T cell receptor gamma variable 9
- Gene Aliases
- TCRGV9, TRGC1, V2
- Chromosome Location
- Chr. 7: 38317017 - 38317492 on Build GRCh38
- UniGene ID
- Hs.534032
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC072387.1 |
- |
3, 4 |
706 |
|
BC072396.1 |
- |
5, 6 |
1132 |
|
M16768.1 |
- |
4, 5 |
892 |
|
View Full Product Details