Gene Transcripts
- Gene Symbol
- TARP
- Entrez Gene ID
- 445347
- Gene Name
- TCR gamma alternate reading frame protein
- Gene Aliases
- CD3G, TCRG, TCRGC1, TCRGC2, TCRGV
- Chromosome Location
- Chr. 7: 38239577 - 38276741 on Build GRCh38
- UniGene ID
- Hs.534032
- Species
- Homo sapiens
- Gene Symbol
- TRGC2
- Entrez Gene ID
- 6967
- Gene Name
- T cell receptor gamma constant 2
- Gene Aliases
- TCRGC2, TRGC2(2X), TRGC2(3X)
- Chromosome Location
- Chr. 7: 38240024 - 38249572 on Build GRCh38
- UniGene ID
- Hs.731375
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
M13231.1 |
- |
7 |
860 |
|
M17323.1 |
- |
3 |
498 |
|
- Gene Symbol
- TRGV9
- Entrez Gene ID
- 6983
- Gene Name
- T cell receptor gamma variable 9
- Gene Aliases
- TCRGV9, TRGC1, V2
- Chromosome Location
- Chr. 7: 38317017 - 38317492 on Build GRCh38
- UniGene ID
- Hs.534032
- Species
- Homo sapiens
View Full Product Details