Gene Transcripts

Gene Symbol
FAAP20
Entrez Gene ID
199990
Gene Name
Fanconi anemia core complex associated protein 20
Gene Aliases
C1orf86, FP7162
Chromosome Location
Chr. 1: 2184460 - 2212720 on Build GRCh38
UniGene ID
Hs.107101
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK054818.1 - 1 1229
AK093927.1 - 1 718
AK125445.1 - 1 2076
AK128507.1 - 1 3064

View Full Product Details