Gene Transcripts
- Gene Symbol
- PHLDB2
- Entrez Gene ID
- 90102
- Gene Name
- pleckstrin homology like domain family B member 2
- Gene Aliases
- LL5b, LL5beta
- Chromosome Location
- Chr. 3: 111732480 - 111976517 on Build GRCh38
- UniGene ID
- Hs.603252
- Species
- Homo sapiens
- Gene Symbol
- LOC100128035
- Entrez Gene ID
- 100128035
- Gene Name
- VGSA5840
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.650705
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AY358772.1 |
- |
1 |
271 |
|
View Full Product Details