Gene Transcripts

Gene Symbol
SV2B
Entrez Gene ID
9899
Gene Name
synaptic vesicle glycoprotein 2B
Gene Aliases
HsT19680
Chromosome Location
Chr. 15: 91099588 - 91301309 on Build GRCh38
UniGene ID
Hs.21754
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001167580.1 NP_001161052.1 12 6481
NM_001323031.1 NP_001309960.1 13 7014
NM_001323032.1 NP_001309961.1 13 7323
NM_001323033.1 NP_001309962.1 11 6990
NM_001323034.1 NP_001309963.1 12 7122
NM_001323036.1 NP_001309965.1 12 6300
NM_001323037.1 NP_001309966.1 14 7230
NM_001323038.1 NP_001309967.1 13 7142
NM_001323039.1 NP_001309968.1 13 7415
NM_001323040.1 NP_001309969.1 13 6661
NM_014848.5 NP_055663.1 14 7411
XM_005254998.3 XP_005255055.1 13 7076
XM_017022761.1 XP_016878250.1 13 7027
XM_017022762.1 XP_016878251.1 13 7162
GenBank mRNA AK124722.1 - 1 1072

View Full Product Details