Gene Transcripts
- Gene Symbol
- TMEM35B
- Entrez Gene ID
- 100506144
- Gene Name
- transmembrane protein 35B
- Gene Aliases
- ZMYM6NB
- Chromosome Location
- Chr. 1: 34981526 - 34985347 on Build GRCh38
- UniGene ID
- Hs.533986
- Species
- Homo sapiens
- Gene Symbol
- ZMYM6
- Entrez Gene ID
- 9204
- Gene Name
- zinc finger MYM-type containing 6
- Gene Aliases
- Buster2, MYM, ZBED7, ZNF198L4, ZNF258
- Chromosome Location
- Chr. 1: 34986165 - 35031968 on Build GRCh38
- UniGene ID
- Hs.533986
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AV734109.1 |
- |
2 |
144 |
|
View Full Product Details