Gene Transcripts

Gene Symbol
KRTAP25-1
Entrez Gene ID
100131902
Gene Name
keratin associated protein 25-1
Gene Aliases
KAP25.1
Chromosome Location
Chr. 21: 30289145 - 30289514 on Build GRCh38
UniGene ID
Hs.580882
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001128598.1 NP_001122070.1 1 167
GenBank mRNA AB180043.1 - 1 167

View Full Product Details