Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001282275.1 | NP_001269204.1 | 2 | 475 | |
NM_001282276.1 | NP_001269205.1 | 2 | 409 | ||
XM_006721865.3 | XP_006721928.1 | 2 | 1091 | ||
XM_011524747.2 | XP_011523049.1 | 2 | 401 | ||
XM_011524748.1 | XP_011523050.1 | 2 | 346 | ||
GenBank mRNA | AI762586.1 | - | 1 | 98 | |
BX106051.1 | - | 2 | 409 |