Gene Transcripts

Gene Symbol
PREB
Entrez Gene ID
10113
Gene Name
prolactin regulatory element binding
Gene Aliases
SEC12
Chromosome Location
Chr. 2: 27130756 - 27134674 on Build GRCh38
UniGene ID
Hs.279784
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013388.4 NP_037520.1 3 709
XM_006711914.2 XP_006711977.1 3 690
XM_011532471.2 XP_011530773.1 3 690
XM_011532472.2 XP_011530774.1 3 690
GenBank mRNA AF203687.1 - 3 587
AF226684.1 - 3 600
AK001976.1 - 1 1423
AK023064.1 - 3 666
AK023405.1 - 3 753
AK292178.1 - 4 596
BC002765.2 - 3 640 3631721
BC012890.1 - 3 650 4158577
BC016472.1 - 3 640 4178911
BC016906.1 - 3 583 4427483
BC041032.1 - 3 703 4638313

View Full Product Details