Gene Transcripts

Gene Symbol
MPHOSPH10
Entrez Gene ID
10199
Gene Name
M-phase phosphoprotein 10
Gene Aliases
CT90, MPP10, MPP10P, PPP1R106
Chromosome Location
Chr. 2: 71130314 - 71150102 on Build GRCh38
UniGene ID
Hs.656208
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005791.2 NP_005782.1 2 636
GenBank mRNA AK056862.1 - 2 636
AK310931.1 - 2 318
BC041084.1 - 2 286
BC126389.1 - 2 573 8992105
X98494.1 - 2 259

View Full Product Details