Gene Transcripts

Gene Symbol
ZSWIM7
Entrez Gene ID
125150
Gene Name
zinc finger SWIM-type containing 7
Gene Aliases
SWS1
Chromosome Location
Chr. 17: 15976560 - 15999692 on Build GRCh38
UniGene ID
Hs.593985
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001042697.1 NP_001036162.1 3 233
NM_001042698.1 NP_001036163.1 3 233
GenBank mRNA BC018807.1 - 3 204
BC044842.1 - 3 191
BC066344.1 - 3 225
BC089386.1 - 3 224
BC094855.1 - 4 428
BU075283.1 - 3 236
DQ530600.1 - 3 135

View Full Product Details