Gene Transcripts

Gene Symbol
SMCR8
Entrez Gene ID
140775
Gene Name
Smith-Magenis syndrome chromosome region, candidate 8
Gene Aliases
Chromosome Location
Chr. 17: 18315280 - 18328056 on Build GRCh38
UniGene ID
Hs.592944
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144775.2 NP_658988.2 1 1582
GenBank mRNA AF467440.1 - 1 1460
AK296847.1 - 1 1570
BC101115.1 - 1 1456
BC101116.2 - 1 1456
BC101117.2 - 1 1456
BC142680.1 - 1 1552 40148405

View Full Product Details