Gene Transcripts

Gene Symbol
PRSS36
Entrez Gene ID
146547
Gene Name
protease, serine 36
Gene Aliases
Chromosome Location
Chr. 16: 31138926 - 31150094 on Build GRCh38
UniGene ID
Hs.256632
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001258290.1 NP_001245219.1 13 1967
NM_001258291.1 NP_001245220.1 13 1982
NM_173502.4 NP_775773.2 13 1982
XM_011545758.2 XP_011544060.1 12 2200
XM_017022968.1 XP_016878457.1 13 1971
XM_017022969.1 XP_016878458.1 12 1829
GenBank mRNA AJ627034.1 - 13 1954
AK075142.1 - 13 1955
AK290310.1 - 13 1982
AK297997.1 - 11 1923
AK301409.1 - 13 1954
BC137396.1 - 13 1949 9021394
BC144615.1 - 13 1934 9053146

View Full Product Details