Gene Transcripts

Gene Symbol
WBSCR27
Entrez Gene ID
155368
Gene Name
Williams Beuren syndrome chromosome region 27
Gene Aliases
Chromosome Location
Chr. 7: 73834590 - 73842527 on Build GRCh38
UniGene ID
Hs.647042
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152559.2 NP_689772.2 6 689
XM_017011777.1 XP_016867266.1 6 769
XM_017011778.1 XP_016867267.1 6 891

View Full Product Details