Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000777.4 | NP_000768.1 | 8 | 844 | |
NM_001291829.1 | NP_001278758.1 | 9 | 1344 | ||
NM_001291830.1 | NP_001278759.1 | 9 | 1104 | ||
NR_033807.2 | 9 | 1363 | |||
XM_006715859.2 | XP_006715922.1 | 8 | 811 | ||
XM_011515843.1 | XP_011514145.1 | 9 | 961 | ||
XM_017011781.1 | XP_016867270.1 | 5 | 486 | ||
XM_017011782.1 | XP_016867271.1 | 3 | 283 | ||
GenBank mRNA | AK223008.1 | - | 8 | 814 | |
AK296205.1 | - | 9 | 1071 | ||
AK309166.1 | - | 10 | 1054 | ||
AK313813.1 | - | 8 | 811 | ||
BC033862.1 | - | 8 | 803 | 5188554 | |
BX537676.1 | - | 9 | 1336 | ||
BX648856.1 | - | 9 | 1315 | ||
J04813.1 | - | 8 | 801 | ||
L26985.1 | - | 10 | 1034 | ||
X90579.1 | - | 9 | 933 |