Gene Transcripts

Gene Symbol
EXD1
Entrez Gene ID
161829
Gene Name
exonuclease 3'-5' domain containing 1
Gene Aliases
EXDL1
Chromosome Location
Chr. 15: 41182728 - 41231074 on Build GRCh38
UniGene ID
Hs.307999
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286441.1 NP_001273370.1 12 1815
NM_152596.3 NP_689809.2 10 1560
XM_006720415.3 XP_006720478.1 11 1732
XM_011521298.2 XP_011519600.1 13 2045
XM_011521299.2 XP_011519601.1 13 1776
XM_011521300.2 XP_011519602.1 10 1304
XM_011521301.2 XP_011519603.1 12 1601
XM_011521302.2 XP_011519604.1 7 1118
XM_017021970.1 XP_016877459.1 10 1304
XM_017021971.1 XP_016877460.1 10 1302
GenBank mRNA AK123414.1 - 2 861
AK292524.1 - 10 1466
AK302864.1 - 12 1749
BC030628.2 - 10 1500 4827307

View Full Product Details