Gene Transcripts

Gene Symbol
CFD
Entrez Gene ID
1675
Gene Name
complement factor D
Gene Aliases
ADIPSIN, ADN, DF, PFD
Chromosome Location
Chr. 19: 859659 - 863569 on Build GRCh38
UniGene ID
Hs.155597
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001317335.1 NP_001304264.1 3 394
NM_001928.3 NP_001919.2 3 373
GenBank mRNA AJ313463.1 - 3 342
AK300963.1 - 3 368
BC034529.1 - 2 289
BC040146.1 - 2 275
BC051001.1 - 3 313
BC057807.1 - 3 372 5269709
CR536575.1 - 2 267
DA008405.1 - 3 390
DQ884409.1 - 3 363
HY087483.1 - 3 374
M84526.1 - 3 321

View Full Product Details