Gene Transcripts

Gene Symbol
UBR1
Entrez Gene ID
197131
Gene Name
ubiquitin protein ligase E3 component n-recognin 1
Gene Aliases
JBS
Chromosome Location
Chr. 15: 42942897 - 43106088 on Build GRCh38
UniGene ID
Hs.591121
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_174916.2 NP_777576.1 44, 45 4909
GenBank mRNA AF525401.1 - 44, 45 4792
AK127478.1 - 8, 9 764
AY061886.1 - 44, 45 4843
BC113505.1 - 44, 45 4921 8322557
BC113507.1 - 44, 45 4921 8322559
BX648400.1 - 16, 17 1721

View Full Product Details