Gene Transcripts
- Gene Symbol
- SLC39A12
- Entrez Gene ID
- 221074
- Gene Name
- solute carrier family 39 member 12
- Gene Aliases
- LZT-Hs8, ZIP-12, bA570F3.1
- Chromosome Location
- Chr. 10: 17951839 - 18043292 on Build GRCh38
- UniGene ID
- Hs.350895
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK055061.1 |
- |
5 |
1087 |
|
View Full Product Details