Gene Transcripts

Gene Symbol
VPS39
Entrez Gene ID
23339
Gene Name
VPS39, HOPS complex subunit
Gene Aliases
TLP, VAM6, hVam6p
Chromosome Location
Chr. 15: 42158701 - 42208331 on Build GRCh38
UniGene ID
Hs.88025
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001301138.1 NP_001288067.1 25 2698
NM_015289.3 NP_056104.2 24 2665
XM_011521403.2 XP_011519705.1 25 2667
XM_011521404.2 XP_011519706.1 24 2670
XM_017022036.1 XP_016877525.1 25 2666
XM_017022037.1 XP_016877526.1 24 2670
XM_017022038.1 XP_016877527.1 24 2853
XM_017022039.1 XP_016877528.1 24 2664
GenBank mRNA AB018313.2 - 24 2616
AF280814.1 - 25 2525
AF281052.1 - 24 2635
AK025334.1 - 9 865
AK128403.1 - 7 1987
BC015817.2 - 20 2225
BC068559.1 - 24 2634 5295166

View Full Product Details