Gene Transcripts
- Gene Symbol
- EHBP1L1
- Entrez Gene ID
- 254102
- Gene Name
- EH domain binding protein 1 like 1
- Gene Aliases
- Chromosome Location
- Chr. 11: 65576034 - 65592650 on Build GRCh38
- UniGene ID
- Hs.502867
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001099409.1 |
NP_001092879.1
|
7, 8 |
969 |
|
XM_005273867.3 |
XP_005273924.1
|
8, 9 |
1008 |
|
XM_005273868.3 |
XP_005273925.1
|
8, 9 |
1007 |
|
XM_005273869.3 |
XP_005273926.1
|
8, 9 |
1009 |
|
XM_005273870.3 |
XP_005273927.1
|
8, 9 |
1008 |
|
XM_006718486.3 |
XP_006718549.1
|
8, 9 |
1006 |
|
XM_006718487.3 |
XP_006718550.1
|
8, 9 |
1006 |
|
XM_017017467.1 |
XP_016872956.1
|
8, 9 |
756 |
|
XM_017017468.1 |
XP_016872957.1
|
8, 9 |
733 |
|
XM_017017469.1 |
XP_016872958.1
|
7, 8 |
971 |
|
GenBank mRNA |
AK024451.1 |
- |
3, 4 |
381 |
|
AL834433.1 |
- |
7, 8 |
666 |
|
View Full Product Details