Gene Transcripts

Gene Symbol
SNX32
Entrez Gene ID
254122
Gene Name
sorting nexin 32
Gene Aliases
SNX6B
Chromosome Location
Chr. 11: 65833636 - 65854687 on Build GRCh38
UniGene ID
Hs.591950
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152760.2 NP_689973.2 9, 10 940
XM_006718488.3 XP_006718551.1 9, 10 1243
XM_017017470.1 XP_016872959.1 9, 10 1218
XM_017017471.1 XP_016872960.1 10, 11 1402
XM_017017472.1 XP_016872961.1 8, 9 1604
XM_017017473.1 XP_016872962.1 8, 9 1604
GenBank mRNA AK055496.1 - 7, 8 1303
AK289587.1 - 9, 10 906
AK299512.1 - 7, 8 934
AL833261.1 - 9, 10 926
BC040981.2 - 9, 10 916 4797976
BC045563.1 - 9, 10 889 4815203

View Full Product Details