Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001164277.1 | NP_001157749.1 | 5 | 1170 | |
NM_001164278.1 | NP_001157750.1 | 5 | 854 | ||
NM_001164279.1 | NP_001157751.1 | 5 | 596 | ||
NM_001164280.1 | NP_001157752.1 | 3 | 612 | ||
NM_001467.5 | NP_001458.1 | 4 | 666 | ||
GenBank mRNA | AB209026.1 | - | 1 | 3023 | |
AF110819.1 | - | 3 | 413 | ||
AF110820.1 | - | 3 | 413 | ||
AF110821.1 | - | 2 | 734 | ||
AF110822.1 | - | 3 | 413 | ||
AF111852.1 | - | 1 | 1166 | ||
AK289642.1 | - | 5 | 1138 | ||
AK294993.1 | - | 5 | 732 | ||
AK298395.1 | - | 5 | 550 | ||
AK300646.1 | - | 5 | 562 | ||
AK312628.1 | - | 5 | 836 | ||
AY423732.1 | - | 3 | 433 | ||
BC002400.2 | - | 3 | 614 | 2822215 | |
BC003589.1 | - | 3 | 614 | 2822215 | |
BC014663.1 | - | 3 | 621 | 4857067 | |
BC015650.2 | - | 4 | 1086 | 4660508 | |
BC064563.1 | - | 3 | 609 | 5580134 | |
CR456733.1 | - | 3 | 413 | ||
Y15409.1 | - | 3 | 582 |