Gene Transcripts

Gene Symbol
LETMD1
Entrez Gene ID
25875
Gene Name
LETM1 domain containing 1
Gene Aliases
1110019O13Rik, HCCR-1, HCCR-2, HCCR1, HCRR-2
Chromosome Location
Chr. 12: 51048230 - 51068984 on Build GRCh38
UniGene ID
Hs.655272
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001243689.1 NP_001230618.1 8 1034
NM_001300765.1 NP_001287694.1 5 623
NM_015416.4 NP_056231.3 8 995
NR_045017.1 8 870
NR_045018.1 7 879
NR_045019.1 6 796
NR_045020.1 4 507
XM_006719335.1 XP_006719398.1 7 817
XM_006719336.1 XP_006719399.1 7 705
XM_006719339.1 XP_006719402.1 8 933
XM_006719340.1 XP_006719403.1 7 754
XM_011538161.2 XP_011536463.1 8 1002
XM_011538164.2 XP_011536466.1 7 852
XM_011538165.2 XP_011536467.1 5 630
XM_011538167.1 XP_011536469.1 3 424
XM_017019151.1 XP_016874640.1 8 1004
XM_017019152.1 XP_016874641.1 8 970
XM_017019153.1 XP_016874642.1 7 854
XM_017019154.1 XP_016874643.1 7 817
XM_017019155.1 XP_016874644.1 8 842
XM_017019156.1 XP_016874645.1 6 702
XM_017019157.1 XP_016874646.1 6 664
XM_017019158.1 XP_016874647.1 7 705
XM_017019159.1 XP_016874648.1 5 633
XM_017019160.1 XP_016874649.1 5 595
XM_017019161.1 XP_016874650.1 5 601
XM_017019162.1 XP_016874651.1 5 571
XM_017019163.1 XP_016874652.1 6 664
XM_017019164.1 XP_016874653.1 7 871
XM_017019165.1 XP_016874654.1 8 933
XM_017019166.1 XP_016874655.1 7 754
GenBank mRNA AF195651.2 - 8 967
AF315598.1 - 8 831
AF329277.1 - 7 878
AK000639.1 - 8 982
AK057866.1 - 8 940
AK123080.1 - 1 581
AK127540.1 - 8 995
AK225836.1 - 8 953
AK301955.1 - 8 870
AK315119.1 - 8 962
AL050286.1 - 3 487
AY259835.1 - 6 738
AY259836.1 - 4 449
BC000395.2 - 3 244
BC019274.1 - 3 244
BC064943.1 - 8 941 6009854
BM549678.1 - 479
BX343550.2 - 6 737
BX376670.2 - 5 564

View Full Product Details