Gene Transcripts

Gene Symbol
CHRDL2
Entrez Gene ID
25884
Gene Name
chordin like 2
Gene Aliases
BNF1, CHL2
Chromosome Location
Chr. 11: 74696428 - 74731385 on Build GRCh38
UniGene ID
Hs.432379
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001278473.2 NP_001265402.1 9 1544
NM_001304390.1 NP_001291319.1 8 1436
NM_001304391.1 NP_001291320.1 7 1241
NM_001304415.1 NP_001291344.1 7 1241
NM_001304416.1 NP_001291345.1 7 1342
NM_001304417.1 NP_001291346.1 6 1134
NM_015424.5 NP_056239.3 9 1544
GenBank mRNA AF332891.1 - 6 825
AK304519.1 - 9 1312
AL047873.1 - 1 29
AL110168.1 - 3 446
AY163868.1 - 9 1050
AY279090.1 - 9 1341
AY279091.1 - 8 1146
AY279092.1 - 7 1342
AY279093.1 - 7 1342
AY279094.1 - 6 1248
AY358522.1 - 9 1333
BC142623.1 - 9 1544 40147467

View Full Product Details