Gene Transcripts
- Gene Symbol
- CHRDL2
- Entrez Gene ID
- 25884
- Gene Name
- chordin like 2
- Gene Aliases
- BNF1, CHL2
- Chromosome Location
- Chr. 11: 74696428 - 74731385 on Build GRCh38
- UniGene ID
- Hs.432379
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001278473.2 |
NP_001265402.1
|
2, 3 |
698 |
|
NM_001304390.1 |
NP_001291319.1
|
1, 2 |
590 |
|
NM_001304391.1 |
NP_001291320.1
|
1, 2 |
590 |
|
NM_001304415.1 |
NP_001291344.1
|
1, 2 |
590 |
|
NM_015424.5 |
NP_056239.3
|
2, 3 |
698 |
|
GenBank mRNA |
AK304519.1 |
- |
2, 3 |
466 |
|
AY163868.1 |
- |
2, 3 |
204 |
|
AY279090.1 |
- |
2, 3 |
495 |
|
AY279091.1 |
- |
2, 3 |
495 |
|
AY279093.1 |
- |
1, 2 |
590 |
|
AY358522.1 |
- |
2, 3 |
487 |
|
BC142623.1 |
- |
2, 3 |
698 |
40147467 |
JN579714.1 |
- |
2, 3 |
201 |
|
View Full Product Details